missing translation for 'onlineSavingsMsg'
Learn More

N-Z-Ethylenediamine hydrochloride, 95%

Product Code. 11915501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11915501 5 g Glass bottle 5g
11341002 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11915501 Supplier Thermo Scientific Acros Supplier No. 439810050
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18807-71-1 | C10H15ClN2O2 | 230.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18807-71-1
Molecular Formula C10H15ClN2O2
Molecular Weight (g/mol) 230.69
MDL Number MFCD00270150
InChI Key QMLKQXIAPAAIEJ-UHFFFAOYSA-N
Synonym benzyl n-2-aminoethyl carbamate hydrochloride, n-cbz-ethylenediamine hydrochloride, benzyl 2-aminoethyl carbamate hydrochloride, n-carbobenzoxy-1,2-diaminoethane hydrochloride, n-z-ethylenediamine hydrochloride, n-benzyloxycarbonyl-1,2-diaminoethane hydrochloride, n-z-1,2-diaminoethane hydrochloride, n-cbz-1,2-diaminoethane hydrochloride, benzyl 2-aminoethyl carbamate hcl, n-benzyloxycarbonylethylenediamine hydrochloride
PubChem CID 12715871
IUPAC Name benzyl N-(2-aminoethyl)carbamate hydrochloride
SMILES Cl.NCCNC(=O)OCC1=CC=CC=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 170°C to 173°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 230.69
Percent Purity 95%
Physical Form Solution
Chemical Name or Material N-Z-Ethylenediamine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • N-Z-Ethylenediamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.