missing translation for 'onlineSavingsMsg'
Learn More

N1-(1-Oxo-2,3-dihydro-1H-inden-5-yl)acetamide, 97%, Thermo Scientific™

Product Code. 10717505
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10717505 25 g 25g
10602853 1 g 1g
10420594 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10717505 Supplier Thermo Scientific Maybridge Supplier No. TL00757EB
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 58161-35-6
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.214
MDL Number MFCD00099465
InChI Key GHUPGGYDRVSZSW-UHFFFAOYSA-N
Synonym n-1-oxo-2,3-dihydro-1h-inden-5-yl acetamide, n1-1-oxo-2,3-dihydro-1h-inden-5-yl acetamide, 5-acetamido-1-indanone, 5-acetylamino-1-indanone, n-1-oxoindan-5-yl acetamide, n-1-oxo-2,3-dihydroinden-5-yl acetamide, acetamide, n-2,3-dihydro-1-oxo-1h-inden-5-yl, 5-acetamido-2,3-dihydro-1-oxo-1h-indene, 5-acetamido-indanone
PubChem CID 312894
IUPAC Name N-(1-oxo-2,3-dihydroinden-5-yl)acetamide
SMILES CC(=O)NC1=CC2=C(C=C1)C(=O)CC2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 189.21
Percent Purity 97%
Chemical Name or Material N1-(1-oxo-2, 3-dihydro-1H-inden-5-yl)acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.