missing translation for 'onlineSavingsMsg'
Learn More

N1-(3-acetyl-4-hydroxyphenyl)acetamide, Thermo Scientific™

Product Code. 10230205
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10230205 5 g 5g
10096412 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10230205 Supplier Thermo Scientific Maybridge Supplier No. BTB10078DE
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7298-67-1
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
InChI Key DIQSYMRVTOVKQT-UHFFFAOYSA-N
Synonym n-3-acetyl-4-hydroxyphenyl acetamide, n1-3-acetyl-4-hydroxyphenyl acetamide, n-3-acetyl-4-hydroxy-phenyl acetamide, 5'-acetamido-2'-hydroxyacetophenone, acetamide, n-3-acetyl-4-hydroxyphenyl, ksc380g7h, 3'-acetyl-4'-hydroxyacetanilide, 5-acetamido-2-hydroxyacetophenone, 5-acetamido-2-hydroxy-acetophenone, n-3-acetyl-4-hydroxyphenyl-acetamide
PubChem CID 81720
IUPAC Name N-(3-acetyl-4-hydroxyphenyl)acetamide
SMILES CC(=O)C1=C(C=CC(=C1)NC(=O)C)O

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 193.2
Percent Purity 97%
Chemical Name or Material N1-(3-Acetyl-4-hydroxyphenyl)acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.