missing translation for 'onlineSavingsMsg'
Learn More

N1-[4-(Trifluoromethoxy)phenyl]-2-chloroacetamide, 97%, Thermo Scientific™

Product Code. 10730314
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10730314 10 g Amber Glass Bottle 10g
10672195 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10730314 Supplier Thermo Scientific Maybridge Supplier No. KM09078EA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 161290-85-3
Molecular Formula C9H7ClF3NO2
Molecular Weight (g/mol) 253.61
MDL Number MFCD00219796
InChI Key MIWYZHHKNDZBCB-UHFFFAOYSA-N
Synonym 2-chloro-n-4-trifluoromethoxy phenyl acetamide, n-chloroacetyl-4-trifluoromethoxy aniline, n1-4-trifluoromethoxy phenyl-2-chloroacetamide, buttpark 30\07-62, acetamide,2-chloro-n-4-trifluoromethoxy phenyl, 2-chloro-4'-trifluoromethoxy acetanilide, pubchem8402, timtec-bb sbb001832
PubChem CID 601031
IUPAC Name 2-chloro-N-[4-(trifluoromethoxy)phenyl]acetamide
SMILES FC(F)(F)OC1=CC=C(NC(=O)CCl)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 253.61
Percent Purity 97%
Chemical Name or Material N1-[4-(Trifluoromethoxy)phenyl]-2-chloroacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.