missing translation for 'onlineSavingsMsg'
Learn More

N1-Methyl-3-chloroaniline, 95%, Thermo Scientific™

Product Code. 10604215
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10604215 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10604215 Supplier Thermo Scientific Maybridge Supplier No. TL00250EA
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 7006-52-2
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C7H8ClN
Linear Formula ClC6H4NHCH3
MDL Number MFCD00052014
Quantity 10 g
Synonym n-methyl-3-chloroaniline, 3-chloro-n-methylbenzenamine, benzenamine, 3-chloro-n-methyl, 3-chloro-phenyl-methyl-amine, n-methyl-m-chloroaniline, n1-methyl-3-chloroaniline, pubchem9202, acmc-209ocl, 3-chloro-1-methylaniline, 3-chloro-n-methyl aniline
InChI Key WFGYSQDPURFIFL-UHFFFAOYSA-N
SMILES CNC1=CC=CC(Cl)=C1
IUPAC Name 3-chloro-N-methylaniline
Molecular Weight (g/mol) 141.60
PubChem CID 138900
Formula Weight 141.6
Percent Purity 95%
Chemical Name or Material N1-methyl-3-chloroaniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.