missing translation for 'onlineSavingsMsg'
Learn More

N1-Methyl-4-(trifluoromethoxy)aniline, 95%, Thermo Scientific™

Product Code. 10486862
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10486862 10 g 10g
10079441 1 g 1g
10016394 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10486862 Supplier Thermo Scientific Maybridge Supplier No. TL00405EA
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 41419-59-4
Molecular Formula C8H8F3NO
Molecular Weight (g/mol) 191.153
MDL Number MFCD00052332
InChI Key MGCCWCLGIPNIBP-UHFFFAOYSA-N
Synonym n-methyl-4-trifluoromethoxy aniline, n1-methyl-4-trifluoromethoxy aniline, benzenamine,n-methyl-4-trifluoromethoxy, methyl 4-trifluoromethoxy phenyl amine, pubchem8511, 4-trifluoromethoxy-n-methylaniline, methyl 4-trifluoromethoxyphenyl amine, methyl-4-trifluoromethoxy-phenyl-amine, n-methyl-4-trifluoromethoxyphenyl amine
PubChem CID 737351
IUPAC Name N-methyl-4-(trifluoromethoxy)aniline
SMILES CNC1=CC=C(C=C1)OC(F)(F)F

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Linear Formula CF3OC6H4NHCH3
Quantity 10 g
Formula Weight 191.15
Percent Purity 95%
Chemical Name or Material N1-methyl-4-(trifluoromethoxy)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.