missing translation for 'onlineSavingsMsg'
Learn More

N1,N1-Dimethyl-1-phenyl-1,2-ethanediamine, 98%

Product Code. 11499430
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499430 250 mg 250mg
11409440 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499430 Supplier Thermo Scientific Alfa Aesar Supplier No. H50291.MD
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6342-21-8 | C10H16N2 | 164.252 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6342-21-8
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.252
MDL Number MFCD01631934
InChI Key NFSAPTWLWWYADB-UHFFFAOYSA-N
Synonym 2-amino-1-phenylethyl dimethylamine, n1,n1-dimethyl-1-phenyl-1,2-ethanediamine, n1,n1-dimethyl-1-phenylethane-1,2-diamine, n*1*,n*1*-dimethyl-1-phenyl-ethane-1,2-diamine, n∼1∼,n∼1∼-dimethyl-1-phenylethane-1,2-diamine, 1,2-ethanediamine, n1,n1-dimethyl-1-phenyl, acmc-20apq4, daa-0-0, n,n-dimethyl-1-phenyl-ethane-1,2-diamine
PubChem CID 241652
IUPAC Name N,N-dimethyl-1-phenylethane-1,2-diamine
SMILES CN(C)C(CN)C1=CC=CC=C1

Specifications

Quantity 250 mg
UN Number UN2735
Formula Weight 164.25
Percent Purity 98%
Chemical Name or Material N1,N1-Dimethyl-1-phenyl-1,2-ethanediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.