missing translation for 'onlineSavingsMsg'
Learn More

Elma™ Lab Clean N10 Concentrated Cleaner

Product Code. 11872681
Change view
Click to view available options
Volume (Metric):
1 L
2.5 L
10 L
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11872681 10 L 10 L
11862681 2.5 L 2.5 L
10411245 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11872681 Supplier Elma™ Supplier No. 8000077
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for use with labware made from metal, including aluminum and light alloys, glass, ceramic or plastic. Elma™ Lab Clean N10 Concentrated Cleaner gently removes emulsions and lime soap deposits and prevents their redeposition.

  • pH-neutral cleaning agent
  • Also a great choice for cleaning glassware
  • Always test the compatibility of Elma Lab Clean N10 with magnesium alloys before use

Specifications

Product Type Lab Clean N10 Cleaner
pH 8.2
Certifications/Compliance Annex III, EC 648/2004
Composition Anionic surfactants 5 to 15%, non-ionic surfactants 5 to 15%, Phosphates <5%, Polycarboxylates &l
Concentration Ultrasonic unit 2%, dishwasher 1%
Container Type Bottle
Content And Storage 5°C to 30°C
For Use With (Application) For dip and splash cleaning of instruments of labs and workshops applications
Volume (Metric) 10 L
compliance-icons
Product Identifier
  • elma lab clean N10 (ELC N10)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: isotridecanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.