missing translation for 'onlineSavingsMsg'
Learn More

N2-Methyl-5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine, 97%, Thermo Scientific™

Product Code. 10602643
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10602643 10 g Amber Glass Bottle 10g
10334242 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10602643 Supplier Thermo Scientific Maybridge Supplier No. CD05460EA
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 25366-22-7
Molecular Formula C4H4F3N3S
Molecular Weight (g/mol) 183.15
MDL Number MFCD00042321
InChI Key ARAFQFMXQDBEPU-UHFFFAOYSA-N
Synonym n-methyl-5-trifluoromethyl-1,3,4-thiadiazol-2-amine, 2-methylamino-5-trifluoromethyl-1,3,4-thiadiazole, n2-methyl-5-trifluoromethyl-1,3,4-thiadiazol-2-amine, 2-methylamino-5-trifluoromethyl-1,3,4-thiadiazol, 1,3,4-thiadiazol-2-amine, n-methyl-5-trifluoromethyl, 1,3,4-thiadiazol-2-amine,n-methyl-5-trifluoromethyl, methyl 5-trifluoromethyl 1,3,4-thiadiazol-2-yl amine, maybridge1_002895, methyl-5-trifluoromethyl-1,3,4 thiadiazol-2-yl-amine
PubChem CID 2775323
IUPAC Name N-methyl-5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine
SMILES CNC1=NN=C(S1)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 183.16
Percent Purity 97%
Chemical Name or Material N2-Methyl-5-(trifluoromethyl)-1, 3, 4-thiadiazol-2-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.