Learn More
R&D Systems™ N21-MAX Vitamin A Free Media Supplement (50X)
Used for controlled neural stem and progenitor cell expansion. R&D Systems N21-MAX Vitamin A Free Media Supplement (50X) is a nutrient-rich, serum-free, and fully defined media formulation.
Brand:  R&D Systems™ AR012
Encompass
Product Code. 15730013
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Alert:
This product contains human transferrin. This transferrin was tested at the donor level using an FDA licensed method and found to be non-reactive for anti-HIV-1/2 and Hepatitis B surface antigen. As no testing can offer complete assurance of freedom from infectious agents, these reagents should be handled as if capable of transmitting infection.
Description
Description
  • Supplied as 50X concentrated solution
  • Limits retinoic acid production to reduce unwanted NSC differentiation
  • Rigorous quality control testing ensures consistency across experiments
  • Eliminates the need for an astrocyte feeder layer
  • Used to mature and maintain healthy primary neurons in culture
  • Quality tested to reduce unwanted experimental variation
  • Contains factors qualified to the expansion of NSCs in vitro
  • Does not require an astrocyte feeder layer
Specifications
Specifications
50X
N21-MAX Vitamin A Free Media Supplement
10 mL
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Albumin (bovine), L-Carnitine, Catalase, Corticosterone, Ethanolamine, Glutathione, Galactose, Holo-Transferrin, Insulin, Linoleic Acid, Linolenic Acid, Lipoic Acid, Progesterone, Putrescine, Selenite, Superoxide dismutase, Triiodo-L-thyronine, D,L-alpha-Tocopherol, D,L-alpha-Tocopherol acetate. Supplied in a volume sufficient to supplement 500mL of media at the recommended concentration. N21-MAX Vitamin A Free Media Supplement is tested for use in Neurobasal Media.