missing translation for 'onlineSavingsMsg'
Learn More

N4-Acetylcytidine-13C5, TRC

Product Code. 30477950
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30477950 10 mg
30481113 1 mg
30479230 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477950 Supplier Toronto Research Chemicals Supplier No. TRCN63304910MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula 13C5 C6 H15 N3 O6
Molecular Weight (g/mol) 290.217
Synonym N-(1,2-Dihydro-2-oxo-1-beta-D-ribofuranosyl-4-pyrimidinyl)acetamide-13C5, 4-Acetylcytidine-13C5, N4-Acetylcytidine-13C5
IUPAC Name N-[1-[(2R,3S,4R,5R)-3,4-dihydroxy-5-(hydroxy(113C)methyl)(2,3,4,5-13C4)oxolan-2-yl]-2-oxopyrimidin-4-yl]acetamide
SMILES CC(=O)NC1=NC(=O)N(C=C1)[13C@@H]2O[13C@H]([13CH2]O)[13C@H](O)[13C@@H]2O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H15N3O6/c1-5(16)12-7-2-3-14(11(19)13-7)10-9(18)8(17)6(4-15)20-10/h2-3,6,8-10,15,17-18H,4H2,1H3,(H,12,13,16,19)/t6-,8+,9+,10-/m1/s1/i4+1,6+1,8+1,9+1,10+1
Formula Weight 290.113
Chemical Name or Material N4-Acetylcytidine-13C5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.