missing translation for 'onlineSavingsMsg'
Learn More

N4-Acetylcytidine triphosphate sodium, MedChemExpress

Product Code. 16495386
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495386 100 mg 100mg
16477247 5 mg 5mg
16470837 50 mg 50mg
16434538 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495386 Supplier Medchem Express Supplier No. HY111815A
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress N4-Acetylcytidine triphosphate sodium is efficiently used as a substrate in T7 Polymerase-catalyzed in vitro transcription and it can be incorporated into multiple templates.

N4-Acetylcytidine triphosphate sodium is efficiently used as a substrate in T7 Polymerase-catalyzed in vitro

Specifications

Chemical Name or Material N4-Acetylcytidine triphosphate sodium
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H14N3Na4O15P3
Quantity 100 mg
Synonym ac4CTP sodium
Solubility Information H2O : ≥ 250 mg/mL (407.75 mM)
SMILES O[C@@H]([C@H]([C@H](N1C(N=C(C=C1)NC(C)=O)=O)O2)O)[C@H]2COP(OP(OP(O[Na])(O[Na])=O)(O[Na])=O)(O[Na])=O
Molecular Weight (g/mol) 613.12
Formula Weight 613.12
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.