missing translation for 'onlineSavingsMsg'
Learn More

N4-Benzoylcytosine, 98%

Product Code. 15509533 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15509533 25 g 25g
15599523 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15509533 Supplier Thermo Scientific Acros Supplier No. 460540250
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 26661-13-2 | C11H9N3O2 | 215.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26661-13-2
Molecular Formula C11H9N3O2
Molecular Weight (g/mol) 215.21
MDL Number MFCD00239434
InChI Key XBDUZBHKKUFFRH-UHFFFAOYSA-N
Synonym n4-benzoylcytosine, n-2-oxo-1,2-dihydropyrimidin-4-yl benzamide, n-benzoylcytosine, n-4-benzoyl cytosine, benzamide, n-1,2-dihydro-2-oxo-4-pyrimidinyl, n-2-oxo-1h-pyrimidin-6-yl benzamide, benzamide, n-2,3-dihydro-2-oxo-4-pyrimidinyl, n-2-oxohydropyrimidin-4-yl benzamide, n-2-oxo-1h-pyrimidin-4-yl benzamide, n-benzoyl cytosine
PubChem CID 309343
IUPAC Name N-(2-oxo-1H-pyrimidin-6-yl)benzamide
SMILES O=C(NC1=CC=NC(=O)N1)C1=CC=CC=C1

Specifications

Melting Point >300.0°C
Elemental Analysis 59.86 to max. 62.93% min. Carbon 19.04 to max. 20.01% min. Nitrogen
Infrared Spectrum Authentic
Quantity 25 g
Formula Weight 215.21
Percent Purity 98%
Chemical Name or Material N4-Benzoylcytosine
compliance-icons
Product Identifier
  • N4-Benzoylcytosine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.