missing translation for 'onlineSavingsMsg'
Learn More

N6-Benzoyl-2'-deoxyadenosine, 98%, Thermo Scientific™

Product Code. 15426249
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15426249 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15426249 Supplier Thermo Scientific Alfa Aesar Supplier No. J64175.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N6-Benzoyl-2′-deoxyadenosine is used in formation of N-Branched oligonucleotides as by-products in solid-phase oligonucleotide synthesis.

Solubility
Soluble in water (slightly), and methanol (20 mg/ml).
TRUSTED_SUSTAINABILITY

Specifications

CAS 4546-72-9
Color White
Molecular Formula C17H17N5O4
MDL Number MFCD00009628
Quantity 5 g
Synonym n6-benzoyl-2'-deoxyadenosine, n-benzoyl-2'-deoxyadenosine, n-benzoyl-2'-deoxy-adenosine, n-9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-yl benzamide, n 6-benzoyl-2'-deoxyadenosine, n6-benzoyl-2-deoxyadenosine, adenosine, n-benzoyl-2'-deoxy, n-benzoyldeoxyadenosine, n6-benzoyldeoxyadenosine, 6-n-benzoyldeoxyadenosine
InChI Key PIXHJAPVPCVZSV-LJEKBPCBNA-N
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12
IUPAC Name N-[9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]purin-6-yl]benzamide
Molecular Weight (g/mol) 355.35
PubChem CID 107558
Formula Weight 355.35
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N6-Benzoyl-2'-deoxyadenosine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.