missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N6-Cyclopentyladenosine, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
N6-Cyclopentyladenosine (CPA) is a selective Adenosine A1 receptor agonist, with Ki values of 2.3 nM, 790 nM and 43 nM for human A1, A2A and A3 receptors, respectively.
Specifications
Specifications
| Chemical Name or Material | N6-Cyclopentyladenosine |
| CAS | 41552-82-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C15H21N5O4 |
| Quantity | 50 mg |
| Synonym | CPA UK-80882 |
| Solubility Information | DMSO : 12.5 mg/mL (37.27 mM; Need ultrasonic) |
| SMILES | OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3NC4CCCC4)O1)O)O |
| Molecular Weight (g/mol) | 335.36 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction