missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N6-Etheno 2'-deoxyadenosine, MedChemExpress
Description
N6-Etheno 2'-deoxyadenosine is a reactive oxygen species (ROS)/reactive nitrogen species (RNS)-induced DNA oxidation product, used as a biomarker to evaluate chronic inflammation and lipid peroxidation in animal or human tissues.
Specifications
Specifications
| Chemical Name or Material | N6-Etheno 2'-deoxyadenosine |
| CAS | 68498-25-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C12H13N5O3 |
| Quantity | 5 mg |
| Solubility Information | H2O : 7.14 mg/mL (25.94 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble) |
| SMILES | O[C@@H](C1)[C@@H](CO)O[C@H]1N(C=N2)C3=C2C4=NC=CN4C=N3 |
| Molecular Weight (g/mol) | 275.26 |
| Formula Weight | 275.26 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?