missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N6-Methyl-2'-deoxyadenosine, 99%

Product Code. 15426669
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426669 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15426669 Supplier Thermo Scientific Chemicals Supplier No. J64961.03
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2002-35-9 | C11H15N5O3 | 265.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2002-35-9
Molecular Formula C11H15N5O3
Molecular Weight (g/mol) 265.27
MDL Number MFCD00055999
InChI Key DYSDOYRQWBDGQQ-XLPZGREQSA-N
PubChem CID 168948
ChEBI CHEBI:7417
SMILES CNC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1

Specifications

Color White
Quantity 1 g
Synonym n6-methyl-2'-deoxyadenosine, n6-me-dado, n 6-me-dado, adenosine, 2'-deoxy-n-methyl, unii-48qzw2ir9h, 2r,3s,5r-2-hydroxymethyl-5-6-methylamino-9h-purin-9-yl tetrahydrofuran-3-ol, n 6-methyl-2'-deoxyadenosine, n6-methyldeoxyadenosine, 2'-deoxy-n6-methylado, 48qzw2ir9h
Solubility Information Soluble in water
IUPAC Name (2R,3S,5R)-2-(hydroxymethyl)-5-[6-(methylamino)purin-9-yl]oxolan-3-ol
Formula Weight 265.27
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N6-Methyl-2'-deoxyadenosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.