Learn More
Macherey-Nagel™ N9 Polypropylene Blue Screw Caps
Shop All Macherey-Nagel Products
Click to view available options
Septum:
Red Rubber/Clear FEP
White Silicone/Blue PTFE
White Silicone/Red PTFE
Unit Size:
Pack of 100
Description
- Screw closures N 9
- Large range of screw closures with different septum materials and cap colors
- Bonded screw caps (septa firmly connected with the cap; cannot be removed) – advantage: thick (blunt) HPLC needles cannot push the septum into the vial
- Polymagnetic screw closure 702166 for CTC GC PAL
Specifications
Specifications
| Color | Blue |
| Material | Polypropylene, PTFE, Silicone |
| Pre-slit | Yes |
| Septum | White Silicone/Blue PTFE |
| Thickness (Metric) | 1 mm |
| Closure Size | N9 |