Learn More
Macherey-Nagel™ N9 Polypropylene Blue Screw Caps
Product Code. 10411708 Shop All Macherey-Nagel Products
Change view
Click to view available options
Septum:
Red Rubber/Clear FEP
White Silicone/Blue PTFE
White Silicone/Red PTFE
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Septum unitSize
10411708 Red Rubber/Clear FEP Pack of 100
10282660 White Silicone/Red PTFE Pack of 100
10327563 White Silicone/Blue PTFE Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10411708 Supplier Macherey-Nagel™ Supplier No. 702732
Only null left
Add to basket
Add to basket
Seal vials with screw caps with a secure septum to receive needles. The Macherey-NagelTM N9 pollypropylene blue screw caps come with a choice of septa.
  • Screw closures N 9
  • Large range of screw closures with different septum materials and cap colors
  • Bonded screw caps (septa firmly connected with the cap; cannot be removed) – advantage: thick (blunt) HPLC needles cannot push the septum into the vial
  • Polymagnetic screw closure 702166 for CTC GC PAL
Specifications
Color Blue
Material FEP, Polypropylene, Rubber
Pre-slit No
Septum Red Rubber/Clear FEP
Thickness (Metric) 1 mm
Closure Size N9