missing translation for 'onlineSavingsMsg'
Learn More

Melford NAD

Product Code. 30018578
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018578 5 g 5g
30018577 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018578 Supplier Melford Supplier No. N301105.0
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

β-Nicotinamide-Adenine Dinucleotide, Oxidized Form (β-NAD) is suitable for Enzyme synthesis / Biocatalysis applications.

β-Nicotinamide-Adenine Dinucleotide, Oxidized Form (β-NAD) is suitable for Enzyme synthesis / Biocatalysis applications. It is also applicable for making Enzyme Multiplied Immunoassay Technique (EMIT), Therapeutic Drug monitoring (TDM) and Blood Glucose reagents.

Specifications

Quantity 5 g
Molecular Formula C21H27N7O14P2 • XH2O
CAS 53-84-9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.