missing translation for 'onlineSavingsMsg'
Learn More

NADH disodium salt, MedChemExpress

Product Code. 16428926
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16428926 100 mg 100mg
16472439 1 g 1g
16442326 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16428926 Supplier Medchem Express Supplier No. HYF0001
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress NADH disodium salt (Disodium NADH) is an orally active reduced coenzyme. NADH disodium salt is a donor of ADP-ribose units in ADP-ribosylaton reactions and a precursor of cyclic ADP-ribose. NADH disodium salt plays a role as a regenerative electron donor in cellular energy metabolism, including glycolysis, β-oxidation and the tricarboxylic acid (TCA) cycle.

NADH disodium salt (Disodium NADH) is an orally active reduced coenzyme. NADH disodium salt is a donor of ADP-ribose units in ADP-ribosylaton reactions and a precursor of cyclic ADP-ribose. NADH disodium salt plays a role as a regenerative electron donor in cellular energy metabolism, including glycolysis, β-oxidation and the tricarboxylic acid (TCA) cycle.

Specifications

Chemical Name or Material NADH disodium salt
CAS 606-68-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H27N7Na2O14P2
Quantity 100 mg
Synonym Disodium NADH
Solubility Information H2O : ≥ 100 mg/mL (140.96 mM) ∣DMSO : 100 mg/mL (140.96 mM; Need ultrasonic)
SMILES NC1=C2C(N([C@H]3[C@H](O)[C@H](O)[C@@H](COP(OP([O-])(OC[C@@H]4[C@@H](O)[C@@H](O)[C@H](N5C=C(C(N)=O)CC=C5)O4)=O)([O-])=O)O3)C=N2)=NC=N1.[Na+].[Na+]
Molecular Weight (g/mol) 709.4
Formula Weight 709.4
Percent Purity 95.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.