missing translation for 'onlineSavingsMsg'
Learn More

Melford NADP

Product Code. 30018567
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018567 100 mg 100mg
30018568 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018567 Supplier Melford Supplier No. N201200.1
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

  • Substance Name: β-Nicotinamide Adenine Dinucleotide Phosphate, Oxidized Form, Monosodium Salt Trihydrate
  • CAS Number: 1184-16-3
  • Molecular Formula: C21H27N7O17P3Na 2H2O
  • Molecular Weight: 801.4 g/mol

Specifications

Quantity 100 mg
Molecular Formula C21H27N7O17P3Na-XH2O
CAS 1184-16-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.