missing translation for 'onlineSavingsMsg'
Learn More

NADP disodium salt, MedChemExpress

Product Code. 16462326
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462326 500 mg 500mg
16448926 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16462326 Supplier Medchem Express Supplier No. HYF0002A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NADP disodium salt (Disodium NADP), a nicotinamide adenine dinucleotide, is a redox cofactor. NADP disodium salt is a key cofactor for electron transfer in the metabolism, being alternately oxidized (NADP+) and reduced (NADPH).

NADP disodium salt (Disodium NADP), a nicotinamide adenine dinucleotide, is a redox cofactor. NADP disodium salt is a key cofactor for electron transfer in the metabolism, being alternately oxidized (NADP+) and reduced (NADPH).

Specifications

Chemical Name or Material NADP disodium salt
CAS 24292-60-2
Color White
Purity Grade Notes Research
Molecular Formula C21H28N7O17P3.2Na
Quantity 500 mg
Synonym Disodium NADP
Solubility Information H2O : 250 mg/mL (316.70 mM; Need ultrasonic) ∣DMSO : 3.33 mg/mL (4.22 mM; ultrasonic and warming and heat to 80°C)
SMILES NC1=C2C(N([C@H]3[C@H](OP(O)(O)=O)[C@H](O)[C@@H](COP(OP([O-])(OC[C@@H]4[C@@H](O)[C@@H](O)[C@H]([N+]5=CC=CC(C(N)=O)=C5)O4)=O)(O)=O)O3)C=N2)=NC=N1.[2 Na]
Molecular Weight (g/mol) 789.38
Formula Weight 789.39
Percent Purity 98.47%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.