missing translation for 'onlineSavingsMsg'
Learn More

NADP sodium salt, MedChemExpress

Product Code. 16452326
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452326 500 mg 500mg
16438926 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16452326 Supplier Medchem Express Supplier No. HYF0002
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NADP sodium salt (Sodium NADP), a nicotinamide adenine dinucleotide, is a redox cofactor. NADP sodium salt is a key cofactor for electron transfer in the metabolism, being alternately oxidized (NADP+) and reduced (NADPH).

NADP sodium salt (Sodium NADP), a nicotinamide adenine dinucleotide, is a redox cofactor. NADP sodium salt is a key cofactor for electron transfer in the metabolism, being alternately oxidized (NADP+) and reduced (NADPH).

Specifications

Chemical Name or Material NADP sodium salt
CAS 1184-16-3
Color White
Purity Grade Notes Research
Molecular Formula C21H27N7NaO17P3
Quantity 500 mg
Synonym Sodium NADP
Solubility Information H2O : 100 mg/mL (130.65 mM; Need ultrasonic) ∣DMSO : 3.57 mg/mL (4.66 mM; ultrasonic and warming and heat to 80°C)
SMILES NC1=C2C(N([C@H]3[C@H](OP(O)(O)=O)[C@H](O)[C@@H](COP(OP([O-])(OC[C@@H]4[C@@H](O)[C@@H](O)[C@H]([N+]5=CC=CC(C(N)=O)=C5)O4)=O)([O-])=O)O3)C=N2)=NC=N1.[Na+]
Molecular Weight (g/mol) 765.39
Formula Weight 765.39
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.