missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford NADPH
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- Substance Name: β-Nicotinamide Adenine Dinucleotide Phosphate, Reduced Form, Tetra Sodium Salt Trihydrate
- CAS Number: 2646-71-1
- Molecular Formula: C21H26N7O17 P3Na4 3H2O
Specifications
Specifications
| Quantity | 500 mg |
| Molecular Formula | C21H26N7O17P3Na4-3H2O |
| CAS | 2646-71-1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?