missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NADPH reduced form, tetrasodium salt
GREENER_CHOICE

Product Code. 18720139
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18720139 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18720139 Supplier Tocris Bioscience™ Supplier No. 5515/100
  • ONLINE EXCLUSIVE PRICE
    331.00 €
    312.90 € / 100mg
    Save 18.10 €
    5% Off
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Cofactor for many anabolic enzymes

NADPH reduced form tetrasodium salt is a cofactor for many anabolic enzymes which use NADPH as a reducing agent.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material NADPH reduced form tetrasodium salt
CAS 2646-71-1
Synonym β-Nicotinamide adenine dinucleotide 2'-phosphate reduced tetrasodium salt
Quantity 100 mg
Solubility Soluble to 50mM in water
Molecular Formula C21H26N7Na4O17P3
Formula Weight 833.35
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >93%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.