missing translation for 'onlineSavingsMsg'
Learn More

Naftidrofuryl oxalate, MedChemExpress

Product Code. 16418616
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16418616 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418616 Supplier Medchem Express Supplier No. HYB1107
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Naftidrofuryl oxalate (Nafronyl oxalate salt) is a drug used in the management of peripheral and cerebral vascular disorders as a vasodilator, enhance cellular oxidative capacity, and may also be a 5-HT2 receptor antagonist.

Naftidrofuryl oxalate (Nafronyl oxalate salt) is a drug used in the management of peripheral and cerebral vascular disorders as a vasodilator, enhance cellular oxidative capacity, and may also be a 5-HT2 receptor antagonist.

Specifications

Chemical Name or Material Naftidrofuryl oxalate
CAS 6-4-3200
Color White
Purity Grade Notes Research
Molecular Formula C26H35NO7
Quantity 100 mg
Synonym Nafronyl oxalate salt
Solubility Information DMSO : ≥ 100 mg/mL (211.17 mM) ∣H2O : 100 mg/mL (211.17 mM; Need ultrasonic)
SMILES O=C(OCCN(CC)CC)C(CC1=C2C=CC=CC2=CC=C1)CC3OCCC3.O=C(O)C(O)=O
Molecular Weight (g/mol) 473.56
Formula Weight 473.56
Percent Purity 96.45%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.