missing translation for 'onlineSavingsMsg'
Learn More

Naftifine Hydrochloride, TRC

Product Code. 30138300
Change view
Click to view available options
Quantity:
50 mg
500 mg
2.5 g
Unit Size:
2.5g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138300 2.5 g 2.5g
30134031 500 mg 500mg
30012029 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138300 Supplier Toronto Research Chemicals Supplier No. TRCN2131002.5G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65473-14-5
Molecular Formula C21 H21 N . Cl H
Molecular Weight (g/mol) 323.86
Synonym N-Methyl-N-[(2E)-3-phenyl-2-propen-1-yl]-1-Naphthalenemethanamine Hydrochloride (1:1), 1-Naphthalenemethanamine, N-methyl-N-(3-phenyl-2-propenyl)-, hydrochloride, (E)-, 1-Naphthalenemethanamine, N-methyl-N-[(2E)-3-phenyl-2-propenyl]-, hydrochloride (9CI), AW 105-843, Exoderil, Naftifine hydrochloride, Naftin, SN 105-843
IUPAC Name (E)-N-methyl-N-(naphthalen-1-ylmethyl)-3-phenylprop-2-en-1-amine;hydrochloride
SMILES Cl.CN(C\C=C\c1ccccc1)Cc2cccc3ccccc23

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C21H21N.ClH/c1-22(16-8-11-18-9-3-2-4-10-18)17-20-14-7-13-19-12-5-6-15-21(19)20;/h2-15H,16-17H2,1H3;1H/b11-8+;
Formula Weight 323.1441
Chemical Name or Material Naftifine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.