missing translation for 'onlineSavingsMsg'
Learn More

Naftopidil hydrochloride, Tocris Bioscience™

Product Code. 30001103
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001103 50 mg 50mg
18712654 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001103 Supplier Tocris Bioscience Supplier No. 0597/50
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

α1 antagonist

An α1-adrenoceptor antagonist with only weak antagonism at post-junctional α2 receptors; a potent, persistent antihypertensive and vasodilator.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1164469-60-6
Molecular Formula C24H29ClN2O3
Molecular Weight (g/mol) 428.957
InChI Key VQAAEWMEVIOHTJ-UHFFFAOYSA-N
Synonym naftopidil hydrochloride, opera_id_518, 1-4-2-methoxyphenyl piperazin-1-yl-3-naphthalen-1-yloxypropan-2-ol hydrochloride, 4-2-methoxyphenyl-?-1-naphthalenyloxy methyl-1-piperazineethanol hydrochloride
PubChem CID 6603044
IUPAC Name 1-[4-(2-methoxyphenyl)piperazin-1-yl]-3-naphthalen-1-yloxypropan-2-ol;hydrochloride
SMILES COC1=CC=CC=C1N2CCN(CC2)CC(COC3=CC=CC4=CC=CC=C43)O.Cl

Specifications

Quantity 50 mg
Formula Weight 428.96
Percent Purity >98%
Chemical Name or Material Naftopidil hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.