missing translation for 'onlineSavingsMsg'
Learn More

Tocris BioScience™ NAIA

Product Code. 31900403
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31900403 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31900403 Supplier Tocris BioScience™ Supplier No. 8910/5
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Broad spectrum cysteine reactive probe; suitable for live-cell applications

NAIA (N-Acryloylindole-alkyne) is a broad-spectrum cysteine reactive probe enabling cysteine profiling and imaging. It covalently binds cysteine residues in proteins.

Displays faster reaction kinetics with CYS (k > 102 M-1 s-1) and targets an expanded pool of ligandable cysteines and proteins compared to IA-Alkyne (Cat. No. 7015). Also shows higher stability and selectivity for CYS over SER and LYS than IA-Alkyne (Cat. No. 7015).

NAIA is used in chemoproteomics experiments as a click-chemistry handle to attach a fluorophore or biotin to reactive cysteines in proteins for visualization by in-gel fluorescence or identification by mass spectrometry, respectively. NAIA can also be used to image oxidized cysteines in cells under oxidative stress by confocal fluorescence microscopy.

Cell-permeable and non-cytotoxic, NAIA is suitable for live-cell experiments.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-5-Hexyn-1-yl-2-[[1-(1-oxo-2-propen-1-yl)-1H-indol-5-yl]oxy]acetamide
CAS 3020685-33-7
Quantity 5 mg
Target Buffers, Chelators and Reagents
Molecular Formula C19H20N2O3
Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.