missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ 500mL LLDPE Safety Bottle Carrier
GREENER_CHOICE

Product Code. 17276020
Change view
Click to view available options
Holds:
Bottles (500 mL)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
17276020 Bottles (500 mL) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17276020 Supplier Thermo Scientific™ Supplier No. 65050010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Secure bottles and reduce chance of bumping, cracking or chipping with this safety bottle carrier.

Secure bottles and reduce chance of bumping, cracking, or chipping. Thermo Scientific™ Nalgene™ 500mL LLDPE Safety Bottle Carrier offers greater rigidity and impact resistance. The sturdy, wide handle is molded in one piece with the base allowing a comfortable grip, even when the carrier is full. This linear low density polyethylene carrier holds up to six standard 500mL reagent bottles in any profile.

  • Cover latches securely to base and allow carriers to stack
  • Dimensions: 14L x 10W x 11 in. H (35 x 25 x 28cm)

Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.

TRUSTED_SUSTAINABILITY

Specifications

Description Carrier Safety Bottle LLDPE 500mL
Color Yellow
Holds Bottles (500 mL)
Material LLDPE
Positions 6
Stackable Yes
No. per Case 6
No. per Pack 1
Product Type Bottle Carrier
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.