missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Copolymer (PPCO) Lowboys with Spigot

Product Code. 10799091
Change view
Click to view available options
Capacity (Metric):
8 L
15 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10799091 8 L Each
10662561 15 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy

Disengage threads and dismount spigot before autoclaving

Follow autoclave instructions provided with the product for proper function

Note: Tubing adapter is not autoclavable

Product Code. 10799091 Supplier Thermo Scientific™ Supplier No. 23240008

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Disengage threads and dismount spigot before autoclaving

Follow autoclave instructions provided with the product for proper function

Note: Tubing adapter is not autoclavable

Dispense most liquids, including reagents, distilled water and titrants, using these polypropylene copolymer (PPCO) lowboys, which fit into tight spaces and can stack three high.

Create a compact, space saving, reagent dispensing bank using chemical-resistant Thermo Scientific™ Nalgene™ Low-Profile autoclavable polypropylene copolymer (PPCO) Lowboys. The low-profile design of these high density polyethylene lowboys fits into tight spaces on shelves and in refrigerators and can stack three high. Chemically compatible with a wide range of lab liquids. Convenient integral carry handle for safe, easy transport. Nalgene polypropylene copolymer carboys can be autoclaved before refilling to eliminate bacterial growth when storing sterile solutions. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.

  • Polypropylene spigot and screw closure
  • Contour handle allows easy one-handed carrying — even when full
  • Can tolerate strong acids and bases for a wide variety of solutions
  • Follow autoclave instructions provided with the product to ensure proper function
  • Autoclavable/Leakproof†

Includes:
Polypropylene screw closures

TRUSTED_SUSTAINABILITY

Specifications

Description Autoclavable Lowboy w/Spigot PP 8 L
Closure Size 63 mm
Type Carboy with Spigot
Material PPCO
Capacity (English) 2 gal.
Capacity (Metric) 8 L
Length (Metric) 305 mm
Diameter (English) Inner Neck 1.73 in.
Diameter (Metric) Inner Neck 44 mm
Width (Metric) 366 mm
Height (English) With Closure 6.26 in.
Height (Metric) With Closure 159 mm
No. per Case 4
No. per Pack 1
Show More Show Less
Do you offer replacement carboy spigots?

The standard Nalgene polyproylene quick action spigot (blue handle and body, white threaded ring for connection to carboy) Cat. No. is 6422-0010 or 6422-0010PK. If the original carboy purchased was a solvent-resistant fluorinated carboy, the replacement (mostly white, with a blue cap on the handle) is the Nalgene Quick-Action Tefzel ETFE Spigot, Cat. No. 6432-0010.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.