missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ HDPE Ultra-Low Particulate Carboy, 20 L, Sterile

Product Code. 16460621
Change view
Click to view available options
Capacity (Metric):
20 L
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
16460621 20 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16460621 Supplier Thermo Scientific™ Supplier No. CEN22890050
  • ONLINE EXCLUSIVE PRICE
    3315.00 €
    2595.00 € / Pack of 4
    Save 720.00 €
    21% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for the most critical applications, Nalgene HDPE Ultra-Low Particulate Bottles and Carboys are platinum certified to contain less than one-third of the USP <788> particulate limits. With exceptionally low particulates and endotoxins, they set the standard for biological and pharmaceutical manufacturing downstream applications.

Our single-use products, including Thermo Scientific™ Nalgene™ HDPE Platinum Certified Clean Bottles and Carboys, help reduce the potential for carry-over contamination. Our new certified platinum clean products include particulate certifications to under 1/3 the allowable limits according to USP<788> and lower endotoxin levels of 0.25 EU/mL, to help mitigate potential sources of unwanted impurities.

  • Products are washed with USP Water for Injection and handled post-washing in Class 5 clean rooms with Class 4 packaging/inspection hoods to produce these very low particulate products
  • Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination
  • Certified low endotoxin (0.25 EU/mL) according to USP <85> to reduce pyrogens
  • Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use
  • Heat-sealed bags include quick-tears on the top and bottom and placed in carton liner twisted and tape/tie sealed to prevent gross contamination on the inner packaging
  • All container/closure systems are certified leakproof* to protect valuable contents
  • Robust, HDPE can be used from -100°C to 120°C, allowing for both heated mixing and frozen storage or shipment. Must be tested in actual use conditions as results may vary depending upon application.
  • Inspection procedures throughout manufacturing include leak testing, dimensional testing, visual inspection for particulates and release testing to ensure the product meets all specifications
  • Products can be supplied with forced extraction studies**, validation binders** and certificates
  • Customer support includes our highly experienced commercial specialist teams, regulatory team, quality team and regional technical support
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Capacity (English) 676.3 oz.
Capacity (Metric) 20 L
Certifications/Compliance < 1/3 particulates allowed according to USP <778>, USP Class VI, non-cytotoxic, USP 30 <661>-compliant, non-pyrogenic, FDA 21 CFR 177.1315
Closure Material Polypropylene
Closure Size 83 mm
Color Clear
Diameter (Metric) Outer 284.2 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Height (English) 19.8 in.
Height (Metric) 501.7 mm
Includes Nalgene Certified Platinum Clean Carboy
Material HDPE
Mouth Style Narrow
No. per Case 4/Cs.
No. per Pack 1 Ea.
Product Type Carboy
Shape Round
Sterility Sterile
Low Particulate <1/3 USP <788> (Ultralow particulate)
Temperature Range (Metric) -100°C to 120°C
Workflow Step Downstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation
Show More Show Less

*Our guarantee for a leakproof seal is subject to our standard product warranty, as set forth in the Thermo Fisher Scientific Terms and Conditions of Sale. Our products are leakproof at ambient temperature and pressure when used with their corresponding closures. However, to ensure safe usage, customers are advised to test our containers and closures under conditions of their planned applications. Please contact technicalsupport@thermofisher.com if you need additional information about our products.

**Forced extraction studies and validation binders from Thermo Fisher Scientific, where available, are provided under a confidentiality agreement to assist customers in product selection. Customers are responsible for determining what studies are recommended for its specific applications. Forced extraction studies and validation binders may be requested by contacting us at ROCregSupport@thermofisher.com.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.