missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Closures for Dropper Bottles: Bulk Pack
GREENER_CHOICE

Product Code. p-4521525
Click to view available options
Color:
Blue
Green
Natural
Red
White
Yellow
missing translation for 'unitSize'
Pack of 2000

Product Code. 10737123

missing translation for 'mfr': Thermo Scientific™ 3127600000

Only null left
Add to basket
Edge
Add to basket

Order these closures, which are available in multiple colors to help identify reagents, to complete your dropper bottle system.

Order Thermo Scientific™ Nalgene™ Closures for Dropper Bottles to complete the Nalgene Dropper Bottle System. Closures fit dropper bottles Cat. Nos. 312750 and 31275, and are available in multiple colors to help identify reagents.

Features:
  • Polypropylene
  • Available in various colors

Ordering Information:

Dropper Bottle Closures are bulk packed 2000 per case. Order Nalgene controlled-drop dispensing tip (Cat. Nos. 312758, 312759) and dropper bottles (Cat. Nos. 312750, 312751) separately.

TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Material Polypropylene
No. per Case 2000
No. per Pack 2000
Neck Finish 15-415
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Dropper Bottle Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Product Type Dropping Bottle Screw Cap
For Use With (Equipment) Dropper Bottles 312750 and 312751.
Closure Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.