Learn More
Thermo Scientific™ Nalgene™ HDPE Quick Disconnects
Product Code. 10345751
Change view
Click to view available options
For Use With:
1/4 to 5/16 in. I.D.
3/8 to 7/16 in. I.D.
Unit Size:
Pack of 12
Pack of 72
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
10345751 1/4 to 5/16 in. I.D. Pack of 12
11374535 3/8 to 7/16 in. I.D. Pack of 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10345751 Supplier Thermo Scientific™ Supplier No. 61500010
  • ONLINE EXCLUSIVE PRICE
    38.44 €
    34.83 € / Pack of 12
    Save 3.61 €
    9% Off
In Stock
Add to basket
Add to basket
Create rapid, smooth connections between two fluid transfer paths with Thermo Scientific™ Nalgene™ Quick Disconnects.
Create rapid, smooth connections between two fluid transfer paths with Thermo Scientific™ Nalgene™ HDPE Quick Disconnects, which allow tubulations to slide together snugly with a twist.
Two tapered tubulations fit tightly together with male/female connection, allowing for rapid joining or separation of apparatus lines.
TRUSTED_SUSTAINABILITY
Specifications
Material HDPE
For Use With 1/4 to 5/16 in. I.D.
Packaging Quantity 12/Pk., 72/Cs.
Product Type Quick Disconnect
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.