missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Large Narrow-Mouth LDPE Bottles
GREENER_CHOICE

Product Code. 10375841
Change view
Click to view available options
Capacity (Metric):
2 L
4 L
8 L
Closure Size:
38-430 mm
53B
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Size unitSize
10375841 2 L 38-430 mm Each
10410723 4 L 38-430 mm Each
10070461 8 L 53B Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10375841 Supplier Thermo Scientific™ Supplier No. 22020005
  • ONLINE EXCLUSIVE PRICE
    14.35 €
    14.05 € / Pack of 1
    Save 0.30 €
    2% Off
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Collect and store distilled water in bottles that offer flexibility, impact resistance and visibility of contents.

Store chemicals and standards and collect and store distilled water. Thermo Scientific™ Nalgene™ Large Narrow-Mouth LDPE Bottles are translucent and offer flexibility, impact resistance and visibility of contents. The extremely low trace metal content in low-density polyethylene makes them ideal for trace metal analysis. These bottles provide excellent chemical resistance to most acids, bases and alcohols. Made only from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables.

Features:
  • Excellent chemical resistance to most acids, bases and alcohols make these versatile bottles
  • Can be used in freezer to -100°C (-148°F) for long-term storage
  • 4 and 8L sizes feature a built-in shoulder loop on larger sizes for attaching identification tag
  • Translucence allows for easy viewing of liquid levels
  • Linerless caps provide the ultimate in leakproof† protection without the use of liner that can wrinkle, cause leaks or contiminate your reagents

Includes:

Leakproof† polypropylene screw caps

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.5 gal., 2 L
Capacity (Metric) 2 L
Capacity (English) 0.5 gal.
Color Translucent
Description Large Narrow-Mouth Bottle
Diameter (Metric) Inner Neck 25 mm
Material LDPE
No. per Case 6
No. per Pack 1
Sterility Non-sterile
Closure Color White
Neck Finish 38 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Mouth Narrow
Shape Round
Height (Metric) 249 mm
Product Type Narrow Mouth Bottle
Height (English) 9.8 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 38-430 mm
Diameter (Metric) 121 mm
Through the Closure Dispensing No
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.