missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box

Product Code. 10185433 Shop All Thermo Scientific Products
Click to view available options
Holds:
Cryogenic Box, 81-Ampule
For Use With (Equipment):
For Locator Jr. Plus
For: Locator 4 Plus
Locator Jr.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10185433

Brand: Thermo Scientific™ HR509X19A70

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Take advantage of efficient, high-volume storage with Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box.

Take advantage of efficient, high-volume storage with Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box.

  • Designed for long-term storage
  • Features an indexed frame and box organizational system
  • Unique hanging rack promotes easy sample retrieval
  • Suited for both manual and computerized inventory record-keeping methods
TRUSTED_SUSTAINABILITY

Specifications

Color Silver
Length (English) 5.25 in.
Width (English) 5.25 in.
For Use With (Equipment) For Locator Jr. Plus
Holds Cryogenic Box, 81-Ampule
Design Locator Jr. Plus
Type Locator™ Plus Cryogenic Rack
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.