missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box
Shop All Thermo Scientific ProductsDescription
Take advantage of efficient, high-volume storage with Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box.
- Designed for long-term storage
- Features an indexed frame and box organizational system
- Unique hanging rack promotes easy sample retrieval
- Suited for both manual and computerized inventory record-keeping methods
Specifications
Specifications
| Color | Gray |
| For Use With (Equipment) | Locator Jr. |
| Holds | Cryogenic Box, 81-Ampule |
| Type | Locator™ Plus Cryogenic Rack |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?