missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Lab Quality Bottles

Product Code. 15528199
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
Closure Size:
13 mm
20 mm
24 mm
28 mm
38-430 mm
Unit Size:
Pack of 12
Pack of 6
Pack of 72
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Size Unit Size
15528199 30 mL 20 mm Pack of 12
30117597 4 mL 13 mm Pack of 12
10142641 8 mL 20 mm Pack of 12
11311584 8 mL 20 mm Pack of 72
15548199 15 mL 20 mm Pack of 12
11361574 60 mL 20 mm Pack of 72
11371574 125 mL 24 mm Pack of 72
10140101 250 mL 24 mm Pack of 12
10281941 500 mL 28 mm Pack of 12
10057960 1 L 38-430 mm Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 15528199 Supplier Thermo Scientific™ Supplier No. 20040001
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Store, ship and package light-sensitive liquids using these narrow-mouth, amber HDPE bottles that offer excellent chemical resistance to most acids, bases and alcohols.

Store, ship and package light-sensitive liquids using Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Lab Quality Bottles. Featuring excellent chemical resistance to most acids, bases and alcohols, these high-density polyethylene bottles are leakproof† when used with appropriate Nalgene closures. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables.

Features:

  • Opaque amber reduces UV light transmissions to protect light-sensitive liquids
  • Meets U.S. Pharmacopoeia 23 requirements for light transmission
  • Versatile bottles where exposure to light needs to be restricted
  • Can be used in freezer to -100°C (-148°F) for long-term storage
  • Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaiminate your reagents

Includes:

Amber polypropylene screw caps

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 1 oz., 30 mL
Capacity (Metric) 30 mL
Color Opaque Amber
Description Lab Quality Amber Narrow-Mouth Bottle
Diameter (Metric) Inner Neck 14 mm
Material HDPE
No. per Case 72
No. per Pack 12
Sterility Non-sterile
Closure Color Amber
Neck Finish 20 mm
Closure Included Yes
Mouth Narrow
Height (Metric) 61 mm
Height (English) 2.4 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 20 mm
Diameter (Metric) 34 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Capacity (English) 1 oz.
Shape Round
Certifications/Compliance Meet U.S. Pharmacopeia XXIII requirements for maximum light transmission.
Product Type Laboratory Bottle
Show More Show Less
Why are your Nalgene containers and caps always different materials?

In order to function as a leakproof system without a liner (one of the key advantages of our laboratory and packaging products), the surfaces of the container and closure that meet and push together when tightened must be dissimilar materials with different hardnesses. Even our polypropylene copolymer (PPCO) bottles with polypropylene homopolymer (PP) bottles are different enough to function properly. On some larger carboy products, this difference cannot be sufficiently maintained so a pliable thermoplastic elastomer (TPE) gasket is added for proper leakproof seal function.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.