missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE IP2 Bottles with Closure: Bulk Pack
BIOPHARMAGREENER_CHOICE

Product Code. 10252673
Change view
Click to view available options
Capacity (Metric):
30 mL
125 mL
250 mL
500 mL
1000 mL
Closure Size:
20-415 mm
24-415 mm
28-415 mm
38-430 mm
Unit Size:
Pack of 1000
Pack of 125
Pack of 250
Pack of 50
Pack of 500
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Size unitSize
10252673 30 mL 20-415 mm Pack of 1000
11329794 125 mL 24-415 mm Pack of 500
10141143 250 mL 24-415 mm Pack of 250
10522045 500 mL 28-415 mm Pack of 125
10272673 1000 mL 38-430 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10252673 Supplier Thermo Scientific™ Supplier No. 3120990001
  • ONLINE EXCLUSIVE PRICE
    1545.00 €
    1501.93 € / Pack of 1000
    Save 43.07 €
    2% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Ship and store liquid reagents using these high density, durable polyethylene bottles with closure. Recommended for customers who design, assemble and certify their own combination packaging.

Ship and store liquid reagents using durable Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE IP2 Bottles with Closure. These high density polyethylene bottles are a safe, economical solution for customers who design, assemble and certify their own combination packaging.

Durable bottles for shipping liquid reagents

  • HDPE bottle with PP closure, bulk pack
  • Narrow mouth
  • Complies with IATA and ICAO regulations

Ordering Information:

Bottles and closures are bulk packed in separate polybags within the same carton

Recommended for:

For storage and shipment of liquid reagents.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 1 oz., 30 mL
Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Translucent
Description IP2 Narrow-Mouth Bottles
Diameter (Metric) Inner Neck 14 mm
Diameter (Metric) Inner 14 mm
For Use With (Application) Storage and Shipment of Liquid Reagents
Height (Metric) With Closure 61 mm
Material HDPE
No. per Case 1000
No. per Pack 1000
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27 (c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9.
Closure Included Yes
Mouth Narrow
Shape Round
Height (Metric) 61 mm
Product Type IP2 Bottle
Height (English) 2.4 in.
Closure Material Polypropylene
Closure Size 20-415 mm
Diameter (Metric) 34 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.