missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure
GREENER_CHOICE

Product Code. 10106120
Change view
Click to view available options
Capacity (Metric):
0.5 L
1 L
2 L
2.5 L
4 L
8 L
Closure Size:
38-430 mm
53B
Unit Size:
Each
Pack of 4
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Size unitSize
10106120 4 L 38-430 mm Each
10060681 0.5 L 38-430 mm Pack of 4
10189630 1 L 38-430 mm Pack of 4
10576922 2 L 38-430 mm Each
10146070 2.5 L 38-430 mm Each
10395841 8 L 53B Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10106120 Supplier Thermo Scientific™ Supplier No. DS22050010
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Freeze samples effectively with impact-resistant these narrow-mouth, polycarbonate bottles with closure.

Freeze samples effectively with impact-resistant Thermo Scientific™ Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure, which are window clear. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables.

Features:

  • 16 oz. and 0.25 gal. sizes are good for smaller volume applications where contents need to be seen
  • 0.5 and 0.66 gal. sizes can be used as culture vessels for batch, suspension or nonattached roller culturing
  • 1 and 2 gal. sizes have built-in shoulder loops for attaching an identification tag
  • Excellent impact resistance makes these bottles good for freezer use to -135°C (-211°F)
  • Linerless caps provide the ultimate in leakproof† protection without the use of liner that can wrinkle, cause leaks or contaminate your reagents
  • Autoclavable/Transparent/Leakproof†

Includes:

Leakproof† polypropylene screw caps

Note:

Before autoclaving, completely disengage threads or remove cap. Autoclave empty. Repeated autoclaving will shorten the bottle life.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 1 gal., 4L
Capacity (Metric) 4 L
Capacity (English) 1 gal.
Color Clear
Description Narrow-Mouth Bottle
Diameter (Metric) Inner Neck 25 mm
Material Polycarbonate
No. per Case 1
No. per Pack 1
Sterility Non-sterile
Closure Color Natural
Neck Finish 38mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1580 for food and beverage use, USP Class VI.
Closure Included Yes
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 333 mm
Product Type Narrow Mouth Bottle
Width (English) 6 in.
Height (English) 13.1 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 38-430 mm
Diameter (Metric) 121 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Autoclavable Autoclavable
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.