missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles with Closure: Bulk Pack
BIOPHARMA

Product Code. p-4521865
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
Neck Finish:
13-415 mm
20-415 mm
Unit Size:
Pack of 2000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Neck Finish unitSize
30117598 4 mL 13-415 mm Pack of 2000
10068872 8 mL 20-415 mm Pack of 2000
10630914 15 mL 20-415 mm Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30117598 Supplier Thermo Scientific™ Supplier No. 3120069125
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Sample, store and ship small-volume, valuable reagents with these autoclavable bottles that can be used in test kits and many small-volume applications.

Sample, store and ship small-volume, valuable reagents. Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles with Closure are durable, autoclavable, and used in test kits and many small-volume applications. These natural, translucent polypropylene copolymer bottles feature uniform walls resistant to splitting or puncturing. Linerless closure, included, ensures performance without wrinkles that can cause leaks or contamination of your solutions.

Features:
  • Nonsterile
  • Autoclavable

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton. Packed 2,000 per case.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.125 oz., 4 mL
Capacity (Metric) 4 mL
Capacity (English) 0.125 oz.
Color Translucent
Diameter (Metric) Inner Neck 8 mm
Material PPCO
No. per Case 2000
No. per Pack 2000
Sterility Non-sterile
Neck Finish 13-415 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Mouth Narrow
Height (Metric) 41 mm
Product Type Sample Container
Height (English) 1.61 in.
Diameter (Metric) 16 mm
Through the Closure Dispensing No
Autoclavable Autoclavable
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.