missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes w/Sealing Cap
GREENER_CHOICE

Product Code. 10088800
Change view
Click to view available options
Capacity (Metric):
10 mL
16 mL
30 mL
50 mL
Length (Metric):
83.8 mm
102.6 mm
107.2 mm
114.8 mm
Diameter (Metric):
16.1 mm
18.2 mm
25.8 mm
28.8 mm
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Length (Metric) Diameter (Metric) unitSize
10088800 10 mL 83.8 mm 16.1 mm Pack of 10
10402121 30 mL 102.6 mm 25.8 mm Pack of 10
10088810 50 mL 114.8 mm 28.8 mm Pack of 10
10179900 16 mL 107.2 mm 18.2 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Autoclaving repeatedly will shorten the life of polycarbonate tubes. Tubes must be filled to at least 80% capacity for proper performance.
Product Code. 10088800 Supplier Thermo Scientific™ Supplier No. 31380010
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Autoclaving repeatedly will shorten the life of polycarbonate tubes. Tubes must be filled to at least 80% capacity for proper performance.

View contents easily using these strong, transparent polycarbonate centrifuge tubes, available with sealing caps for use up to 50,000 x g.

Easily view contents using strong, transparent Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes, equipped with sealing caps for leakproof† performance up to 50,000 x g.

Features:
  • Withstand temperatures from -135° to +135°C (-211° to +275°F)
  • Round bottom to fit high speed rotors
  • Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges
  • Come with PP sealing caps with silicone gaskets for extra leak protection when spinning >10,000 x g or spinning hazardous materials
  • Autoclavable; transparent

Ordering Information:

Sealing caps also sold separately for 30 and 50mL sizes.

TRUSTED_SUSTAINABILITY

Specifications

Closure Material Polypropylene, red rubber
Capacity (Metric) 10 mL
Material Polycarbonate
No. per Case 50
No. per Pack 10
Sterility Non-sterile
Autoclavable Autoclavable
Shape Round
Length (Metric) 83.8 mm
Bottom Shape Round
Closure Type Screw Cap
Writing Area No
Diameter (Metric) 16.1 mm
Graduated No
Product Type Centrifuge Tube
Max. RCF 50,000 x g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.