missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles

Product Code. 12059987
Click to view available options
Capacity (Metric):
125 mL
500 mL
1 L
2 L
5 L
Product Type:
Square Bottle
Quantity:
14 x 5 Bottles
20 Bottles
20 x 5 Bottles
35 Bottles
4 x 5 Bottles
6 Bottles
7 x 5 Bottles
70 Bottles
Unit Size:
Pack of 20
Pack of 35
Pack of 5
Pack of 6
Pack of 70
This item is not returnable. View return policy

Product Code. 12059987

Brand: Thermo Scientific™ 311042

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Eliminate the cost of cleaning, sterilizing and associated validations compared to bottles designed for reuse with Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles, ideal for prolonged storage of pH sensitive materials such as culture media.

Eliminate the cost of cleaning, sterilizing and associated validations compared to bottles designed for reuse with Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles, ideal for prolonged storage of pH sensitive materials such as culture media. Certified sterile and ready-to-use, all Biotainer bottles are gamma irradiated to a Sterility Assurance Level (SAL) of 10–6.

  • Durable, crystal clear PETG for sample integrity
  • Closures with polyethylene foam core liners for leakproof† assurance
  • Custom options available for fluid transfer applications
  • Supported by extensive validation documentation
  • Suitable for storage in conditions from -40° to +70°C

Warranty: 90 days

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1 L
Capacity (English) 33.8 oz.
Color Clear
Diameter (English) 3.8 in.
Diameter (English) Inner Neck 1.46 in.
Diameter (Metric) Inner Neck 37 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​
Material PET
No. per Case 35/Cs.
No. per Pack 5/Pk.
Quantity 7 x 5 Bottles
Sterility Sterile
Temperature Range (Metric) -70°C to 70°C
Certifications/Compliance USP Class VI, Non-cytotoxic, USP <661>, Non-pyrogenic, FDA 21 CFR 177.1315
Closure Included Yes
Graduated Yes
Mouth Narrow
Shape Square
Height (Metric) 201 mm
Product Type Square Bottle
Height (English) 7.9 in.
Closure Liner Polyethylene Foam Core
Closure Size 48 mm
Diameter (Metric) 98 mm
Autoclavable Not Autoclavable
Workflow Step Upstream process solution storage. Upstream Validation
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.