missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Certified Clean Containers, 500mL

Product Code. 10506165
Change view
Click to view available options
:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10506165 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10506165 Supplier Thermo Scientific™ Supplier No. 3820190500x

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific™ Nalgene™ PETG Certified Clean Containers. 

Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific™ Nalgene™ PETG Certified Clean Containers. Suitable for pharmaceutical and biotechnology applications that require processing and storing bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations. A safe alternative for your lab with no sacrifice of accuracy.

Lot-certified for low particulates, these containers features a unique leakproof† design and are suitable for transport.
  • Exhibit excellent gas barrier propertiesand ensure pH stability for buffered solutions.
  • Resins are free of animal-derived components.
  • Sterile, tamper-resistant seal ensures product integrity prior to use.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Capacity (English) 16.9 oz.
Diameter (English) 2.9 in.
Diameter (English) Inner Neck 1.1 in.
Diameter (Metric) Inner Neck 28 mm
Material PETG
No. per Case 24
No. per Pack 4
Sterility Sterile
Certifications/Compliance USP Class VI, European Pharmacopeia for Abnormal Toxicity and USP 661 physicochemical tests compliant. Manufactured in an ISO 14644-1 Class 7 cleanroom. Certified sterile (SAL 10-6), noncytotoxic, nonpyrogenic, and nonhemolytic.
Closure Included Yes
Graduated Yes
Mouth Narrow
Shape Square
Height (Metric) 177 mm
Product Type Clean Container
Height (English) 6.97 in.
Closure Size 38-430 mm
Diameter (Metric) 74 mm
Show More Show Less

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.