missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules
BIOPHARMA

Product Code. 10455123
Change view
Click to view available options
Volume (Metric):
5 mL
10 mL
20 mL
Unit Size:
Pack of 1260
Pack of 1656
Pack of 612
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10455123 5 mL Pack of 1656
10740005 10 mL Pack of 1260
10600334 20 mL Pack of 612
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10455123 Supplier Thermo Scientific™ Supplier No. 3420320005
  • ONLINE EXCLUSIVE PRICE
    937.00 €
    847.00 € / Pack of 1656
    Save 90.00 €
    9% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protect against leakage, breakage and contamination with these PETG serum vials. Lighter in weight than glass, they are made for use with lined continuous-thread closures (available separately).

Protect against leakage, breakage, and contamination with Thermo Scientific™ Nalgene PETG Serum Vials. These serum vials are lighter in weight than glass, and are made for use with lined continuous-thread closures (available separately).

Features:

  • PETG
  • Noncytotoxic
  • Nonpyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Sterile to 10-6 SAL
  • Leakproof† with Nalgene lined continuous-thread closures (Cat. No. 342158)

Ordering Information:

Serum vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 342158).

TRUSTED_SUSTAINABILITY

Specifications

Description Serum Vials
Diameter (Metric) Inner 11 mm
Color Clear
Diameter (Metric) Outer 22 mm
Material PETG
Sterility Sterile
Volume (Metric) 5 mL
No. per Case 1656
No. per Pack 276
Volume (English) 1/6 oz.
Product Line Nalgene
Volume (Metric) Total 5 mL
Closure Type Lined Continuous-thread Closure (Sold Separately)
Quantity 1656 Cases, 276 Pk.
Closure Size 20-415
For Use With (Equipment) Lined Continuous-Thread Closure
Certifications/Compliance ISO 13485:2016
Height (Metric) 44 mm
Product Type Serum Vial
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.