missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure
Perform titrations, preparation and storage of culture media with these flasks, which provide a combination of chemical resistance, strength and transparency.
83.50 € - 132.00 €
Specifications
Material | PMP |
---|---|
Autoclavable | Autoclavable |
Disposable | Reusable |
Graduated | Yes |
Mouth Style | Narrow |
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
10098280
|
Thermo Scientific™
4109-0125 |
125 mL | 33 mm |
83.50 €
Pack of 6 |
In Stock
Log in to see stock. |
||||
10251761
|
Thermo Scientific™
4109-0250 |
250 mL | 38 mm |
86.05 €
Pack of 4 |
In Stock
Log in to see stock. |
||||
10508311
|
Thermo Scientific™
4109-0500 |
500 mL | 43 mm |
132.00 €
Pack of 4 |
In Stock
Log in to see stock. |
||||
10373361
|
Thermo Scientific™
4109-1000 |
1000 mL | 53 mm |
93.65 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
Description
Perform titrations, preparation and storage of culture media with Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure, which provide a combination of chemical resistance, strength and transparency. Rugged, shatter-proof construction combined with light weight make these flasks a valuble element of lab safety programs.
Features:- Excellent chemical resistance to a wide variety of lab reagents
- More transparent than polypropylene but with similar chemical resistance
- Ideal for preparation and storage of culture media and titrations
- All sizes have easy-to-read graduations
- Autoclavable/Graduated
Includes:Polypropylene screw closures
.
Specifications
PMP | |
Reusable | |
Narrow | |
Plain |
Autoclavable | |
Yes | |
Non-sterile | |
Screw |