missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polycarbonate Biotainer™ Bottles and Carboys

Product Code. 10084597
Change view
Click to view available options
Capacity (Metric):
5 mL
20 mL
125 mL
1 L
2 L
5 L
10 L
20 L
Shape:
Round
Square
Square with Handle
Closure Size:
20 mm
38 mm
48 mm
Unit Size:
Pack of 2
Pack of 20
Pack of 3
Pack of 35
Pack of 50
Pack of 500
Pack of 6
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Shape Closure Size Unit Size
10084597 1 L Square 48 mm Pack of 35
10778658 5 mL Round 20 mm Pack of 500
16449469 5 mL Round 20 mm Pack of 500
12438887 20 mL Round 20 mm Pack of 500
10592498 125 mL Square 38 mm Pack of 50
16582380 1 L Square 48 mm -
10432478 2 L Square 48 mm Pack of 20
11724733 5 L Square 48 mm Pack of 6
15941621 5 L Square with Handle 48 mm Pack of 6
16416519 5 L Square with Handle 48 mm Pack of 6
10309347 5 L Square with Handle 48 mm Pack of 6
11859570 10 L Square 48 mm Pack of 2
15951621 10 L Square with Handle 48 mm Pack of 2
10738658 10 L Square with Handle 48 mm Pack of 2
10770160 20 L Square 48 mm Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 10084597 Supplier Thermo Scientific™ Supplier No. 312042
Upgrade Your Bioprocess Workflow
Use promo code 27948
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Nalgene™ Biotainer™ Bottles and Carboys are sterile, ready-to-use and designed specifically for pharmaceutical and biological manufacturers. Excellent for freezing applications.

Reduce the risk of carry-over contamination and eliminate the cost of cleaning, sterilizing and associated validations. Use sterile, ready-to-use Thermo Scientific™ Nalgene™ Biotainer™ Bottles and Carboys, designed specifically for pharmaceutical and biological manufacturers. Excellent for freezing applications.

  • Durable, translucent PC construction ensures the integrity of your sample
  • Closures feature a silicone liner for leakproof† assurance
  • Customization options are available for fluid transfer applications
  • Extensive validation documentation support
  • Suitable for use in storage conditions of -100° to +100°C
  • Not for use on hot plates

Warranty: 90 days

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1 L
Color Clear
Diameter (English) Inner Neck 1.89 in.
Diameter (Metric) Inner Neck 48 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​
Material Polycarbonate
No. per Case 35/Cs.
Sterility Sterile
Temperature Range (Metric) -100°C to 100°C
Liner Material Silicone
Closure Included Yes
Graduated Yes
Mouth Narrow
Height (Metric) 201 mm
Height (English) 7.9 in.
Closure Material Polypropylene
Closure Size 48 mm
Diameter (Metric) 98 mm
Autoclavable Not Autoclavable
Workflow Step Upstream process solution storage. Upstream Validation
Capacity (English) 33.8 oz.
Shape Square
Certifications/Compliance USP Class VI, Non-cytotoxic, USP <661>, Non-pyrogenic
Product Type Storage Bottle
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.