missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polycarbonate Centrifuge Bottles
GREENER_CHOICE

Product Code. p-4521563
Click to view available options
Nominal Capacity:
250 mL
500 mL
1000 mL
For Use With (Equipment):
Fiberlite™ rotors
IEC rotors
missing translation for 'unitSize'
Pack of 16
Pack of 4
This item is not returnable. View return policy

Product Code. 10589291

missing translation for 'mfr': Thermo Scientific™ 31220250

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Process large-volume cell harvesting, pelleting and protein purification with these polycarbonate centrifuge bottles.

Process large-volume cell harvesting, pelleting and protein purification with Thermo Scientific™ Nalgene™ Polycarbonate Centrifuge Bottles, the perfect fit with Thermo Scientific™ Fiberlite™ carbon fiber rotors. Molded of transparent polycarbonate, these clear bottles are autoclavable and have excellent mechanical strength properties.

Molded of transparent polycarbonate for easy visualization of samples, these bottles are ideal for low-speed centrifugation of biological and chemical samples.

  • For added leakproof† assurance at maximum speeds or when spinning hazardous materials, use with polypropylene sealing caps assemblies; available for 250mL (DS3131-0038) and 1000mL (DS3132-0063) sizes
  • Cat. No. 3122-1010 designed for use with IEC rotors
  • For proper performance, bottles must be filled to at least 80% of total capacity

Ordering Information:
When spinning a 250mL bottle in a Thermo Scientific Sorvall GSA rotor, the use of the Nalgene centrifuge bottle adaptor (DS3125-0250) is required. Cat. No. 3122-1010 is for use with IEC Rotors.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Color Clear
Description Polycarbonate Centrifuge Bottles
Material Polycarbonate
No. per Case 36
No. per Pack 4
Nominal Capacity 250 mL
Graduated No
Max. Force 27,500 x g
Shape Round
Height (Metric) 125.2 mm
Product Type Centrifuge Bottle
For Use With (Equipment) Fiberlite™ rotors
Closure Material Polypropylene
Diameter (Metric) 61.8 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Autoclavable Autoclavable
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.