Learn More
Thermo Scientific™ Nalgene™ Polycarbonate Conical-Bottom Centrifuge Bottle Adapter
Product Code. 10788851
Change view
Click to view available options
No. per Pack:
4
Unit Size:
Pack of 4
Product Code. No. per Pack Unit Size
10788851 4 Pack of 4
1 options
Product Code. 10788851 Supplier Thermo Scientific™ Supplier No. DS31260175
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Adapt Thermo Scientific™ Nalgene™ centrifuge bottles Cat. Nos. 3143, 3144 and 3145 for use in a conical-bottom centrifuge with this adapter.
For use with Nalgene 3143-0175 polypropylene copolymer centrifuge bottles, 3144-0175 polycarbonate conical centrifuge bottles, and 3145-0175 polystyrene conical centrifuge bottles.
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Material Polycarbonate
Description Conical Bottom Centrifuge Bottle Adapter
Color White
No. per Pack 4